- [Improving memory effects of eel oil capsule on memory obstruction of mice hurt by anisodine].
[Improving memory effects of eel oil capsule on memory obstruction of mice hurt by anisodine].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2003-02-11)
R He, X Mei, M Gao, D Xu, S Xu
PMID12575128
ABSTRACT
The effects of eel oil capsule on memory obstruction of mice were observed in step-down test, step-through test, Y-maze test and Japanese labyrinth test. The results indicated that the eel oil capsule treating group with dosages of 0.234, 0.702 and 2.106 g/kg markedly enhance the acquirement, strengthing and reappearance of memory, having the functions of improving brain memory hurt by Anisodine.