MilliporeSigma
  • Purification and characterization of a collagenase from the mackerel, Scomber japonicus.
Purification and characterization of a collagenase from the mackerel, Scomber japonicus.
Journal of biochemistry and molecular biology (2002-12-10)
Pyo-Jam Park, Sang-Hoon Lee, Hee-Guk Byun, Soo-Hyun Kim, Se-Kwon Kim
ABSTRACT
Collagenase from the internal organs of a mackerel was purified using acetone precipitation, ion-exchange chromatography on a DEAE-Sephadex A-50, gel filtration chromatography on a Sephadex G-100, ion-exchange chromatography on DEAE-Sephacel, and gel filtration chromatography on a Sephadex G-75 column. The molecular mass of the purified enzyme was estimated to be 14.8 kDa by gel filtration and SDS-PAGE. The purification and yield were 39.5-fold and 0.1% when compared to those in the starting-crude extract. The optimum pH and temperature for the enzyme activity were around pH 7.5 and 55 degrees, respectively. The K(m) and V(max) of the enzyme for collagen Type I were approximately 1.1mM and 2,343 U, respectively. The purified enzyme was strongly inhibited by Hg2+, Zn2+, PMSF, TLCK, and the soybean-trypsin inhibitor.
Sigma-Aldrich
p-Toluenesulfonyl fluoride, 98%