MilliporeSigma
  • Synthesis of the pro-gly dipeptide alkene isostere using olefin cross-metathesis.
Synthesis of the pro-gly dipeptide alkene isostere using olefin cross-metathesis.
The Journal of organic chemistry (2002-08-17)
Melissa M Vasbinder, Scott J Miller
ABSTRACT
An approach to the synthesis of dipeptide olefin isosteres using intermolecular olefin cross-metathesis is presented. In particular, a synthesis of the Pro-Gly isostere (1) is reported. Conversion of N-BOC-proline into the corresponding vinyl-substituted carbamate provides the N-terminal cross-metathesis partner (2). Methyl 3-butenoate (3) is employed as the C-terminal component. Treatment of the two partners in an optimized molar ratio affords the cross product 1 (83% yield). Three other examples are demonstrated to evaluate the potential of the approach.
Sigma-Aldrich
Methyl 3-butenoate, 95%
Sigma-Aldrich
Pro-Gly