MilliporeSigma
  • Adenosine A1 receptor agonists inhibit trigeminovascular nociceptive transmission.
Adenosine A1 receptor agonists inhibit trigeminovascular nociceptive transmission.
Brain : a journal of neurology (2002-05-23)
P J Goadsby, K L Hoskin, R J Storer, L Edvinsson, H E Connor
ABSTRACT
There is a considerable literature to suggest that adenosine A1 receptor agonists may have anti-nociceptive effects, and we sought to explore the role of adenosine A1 receptors in a model of trigeminovascular nociceptive transmission. Cats were anaesthetized (alpha-chloralose 60 mg/kg, intraperitoneally), and prepared for physiological monitoring. The superior sagittal sinus (SSS) was stimulated electrically, and linked units were recorded in the trigeminocervical complex. Post-stimulus histograms were constructed to analyse the responses and the effect of drug administration. Blood was sampled from the external jugular vein to determine levels of calcitonin gene-related peptide (CGRP) release before and after drug administration. Intravenous administration of the highly selective adenosine A1 receptor agonist, GR79236 (3-100 microg/kg) had a dose-dependent inhibitory effect on SSS-evoked trigeminal activity. The maximal effect (80 +/- 6% reduction in probability of firing) was seen at 100 microg/kg. The neuronal inhibitory effect of GR79236 could be inhibited by the selective adenosine A1 receptor antagonist DPCPX (300 microg/kg; P < 0.05). SSS stimulation increased cranial CGRP levels from 33 +/- 2 pmol/l (n = 6) to 64 +/- 3 pmol/l, an effect substantially reduced by pre-treatment with GR79236 (30 microg/kg; P < 0.01). The selective low efficacy adenosine A1 receptor agonist, GR190178 (30-1000 microg/kg i.v.), also inhibited SSS-evoked neuronal activity in a dose-dependent fashion. In this model of trigeminovascular nociception, adenosine A1 receptor activation leads to neuronal inhibition without concomitant vasoconstriction, suggesting a novel avenue for the treatment of migraine and cluster headache.