MilliporeSigma
  • Deoxyartemisinin derivatives from photooxygenation of anhydrodeoxydihydroartemisinin and their cytotoxic evaluation.
Deoxyartemisinin derivatives from photooxygenation of anhydrodeoxydihydroartemisinin and their cytotoxic evaluation.
Journal of natural products (2002-02-23)
Ahmed M Galal, Samir A Ross, Mahmoud A ElSohly, Hala N ElSohly, Farouk S El-Feraly, Mohamed S Ahmed, Andrew T McPhail
ABSTRACT
Photooxygenation of anhydrodeoxydihydroartemisinin (4) followed by chromatographic separation of the reaction mixture yielded the new compounds alpha- (5) and beta-hydroperoxydeoxyartemisitene (8) and the formate ester 7, together with two previously reported compounds, 6 and 9. Reduction of 5 using polymer-bound triphenylphosphine afforded the new compound dihydrodeoxyartemisitene (10). Treatment of 10 with a catalytic amount of BF(3)-OEt(2) yielded the C(2)-symmetrical dimer bis(dihydrodeoxyartemisitene) ether (11) and two new compounds, dihydrodeoxyartemisitene methyl ether (12) and the dimer 13, as minor products. Dehydroacetoxylation of 5 using acetic anhydride in pyridine afforded deoxyartemisitene (14). The identities of the new compounds (5, 7, 8, 10-14) were deduced from their spectral data and by chemical derivatization. The stereochemistry of dimer 11 was defined on the basis of X-ray crystallographic analysis. All compounds were evaluated in vitro in the National Cancer Institute drug-screening program consisting of 60 human cancer cell lines derived from nine different tissues. Of the compounds tested, deoxyartemisitene (14) demonstrated significant cytotoxicity against a number of human cancer cell lines.
Sigma-Aldrich
(3R,3aS,6R,6aS,9S,10aS,10bR)-Octahydro-3,6,9-trimethyl-10aH-9,10b-epoxypyrano[4,3,2-jk][2]benzoxepin-2(3H)-one, AldrichCPR