MilliporeSigma
  • 4-[18F]fluoroarylalkylethers via an improved synthesis of n.c.a. 4-[18F]fluorophenol.
4-[18F]fluoroarylalkylethers via an improved synthesis of n.c.a. 4-[18F]fluorophenol.
Nuclear medicine and biology (2002-02-02)
Thomas Ludwig, Johannes Ermert, Heinz H Coenen
ABSTRACT
This paper describes the improved synthesis of n.c.a. 4- 18F]fluorophenol for the preparation of 18F-labeled alkylarylethers. Nucleophilic fluorination of substituted benzophenone derivatives yielded n.c.a. 4- 18F]fluoro-4'-substituted benzophenones with 80- 90% RCY, which were converted to benzoic acid phenylesters by treatment with peracetic acid. Strong electron-withdrawing substituents like nitro, cyano and trifluoromethyl favor a fluorophenyl-to-oxygen migration resulting in the formation of corresponding benzoic acid fluorophenylesters. N.c.a. 18F]fluorophenol is almost quantitatively formed after hydrolysis and can easily be converted with alkylhalides into n.c.a. 18F]fluoroarylalkylethers.
Sigma-Aldrich
4-Fluorophenol, 99%