MilliporeSigma
  • Serotonin 5-HT2 receptors in schizophrenic patients studied by positron emission tomography.
Serotonin 5-HT2 receptors in schizophrenic patients studied by positron emission tomography.
Life sciences (2000-07-14)
Y Okubo, T Suhara, K Suzuki, K Kobayashi, O Inoue, O Terasaki, Y Someya, T Sassa, Y Sudo, E Matsushima, M Iyo, Y Tateno, M Toru
ABSTRACT
Using positron emission tomography (PET) and [11C]N-methylspiperone (NMSP), we examined 5-HT2 receptors in the cortex of schizophrenic patients in whom we previously observed decreased prefrontal D1 receptor binding. The subjects were 10 neuroleptic-naive schizophrenic patients, 7 schizophrenic patients who were drug-free but had previously been treated with neuroleptics, and 12 normal controls. A non-significant trend towards decreased prefrontal [11C]NMSP binding was observed in the neuroleptic-treated patients, suggesting a possible effect of previous neuroleptic treatment on the alteration in cortical 5-HT2 function. However, the neuroleptic-naive patients showed no noticeable difference in cortical [11C]NMSP binding compared to controls. Our results do not rule out the role of 5-HT2 function as a crucial site of therapeutic activity of schizophrenia, but they do suggest that cortical 5-HT2 receptors might not be primarily involved in the pathophysiology of schizophrenia.