MilliporeSigma
  • Synthesis and biological evaluation of 1,3-oxathiolane 5-azapyrimidine, 6-azapyrimidine, and fluorosubstituted 3-deazapyrimidine nucleosides.
Synthesis and biological evaluation of 1,3-oxathiolane 5-azapyrimidine, 6-azapyrimidine, and fluorosubstituted 3-deazapyrimidine nucleosides.
Nucleosides, nucleotides & nucleic acids (2000-06-08)
M C Liu, M Z Luo, D E Mozdziesz, T S Lin, G E Dutschman, E A Gullen, Y C Cheng, A C Sartorelli
ABSTRACT
(2R,5S)-5-Amino-2-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]- 1,2,4-triazine-3(2H)-one (8) and (2R,5R)-5-amino-2-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-1,2,4-tr iazine-3(2H)-one (9) have been synthesized via a multi-step procedure from 6-azauridine. (2R,5S)-4-Amino-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-1,3, 5-triazine-2(1H)-one (11) and (2R,5R)-4-amino-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]- 1,3,5-triazine-2(1H)-one (12), and the fluorosubstituted 3-deazanucleosides (19-24) have been synthesized by the transglycosylation of (2R,5S)-1-[2-[[(tert-butyldiphenylsilyl) oxy]methyl]-1,3-oxathiolan-5-yl] cytosine (2) with silylated 5-azacytosine and the corresponding silylated fluorosubstituted 3-deazacytosines, respectively, in the presence of trimethylsilyl trifluoromethanesulfonate as the catalyst in anhydrous dichloroethane, followed by deprotection of the blocking groups. These compounds were tested in vitro for cytotoxicity against L1210, B16F10, and CCRF-CEM tumor cell lines and for antiviral activity against HIV-1 and HBV.
Sigma-Aldrich
6-Azauridine