MilliporeSigma
A method for the rapid preparation of 5-vinyluracil in high yield.
Nucleic acids research (1974-01-01)
A S Jones, G P Stephenson, R T Walker
ABSTRACT
A method for the rapid preparation of the thymineanalogue, 5-vinyluracil, in 83% yield from 5-(1-hydroxyethyl)uracil via the methanesulphonyl ester is reported.