- Fast inactivation of a brain K+ channel composed of Kv1.1 and Kvbeta1.1 subunits modulated by G protein beta gamma subunits.
Fast inactivation of a brain K+ channel composed of Kv1.1 and Kvbeta1.1 subunits modulated by G protein beta gamma subunits.
Modulation of A-type voltage-gated K+ channels can produce plastic changes in neuronal signaling. It was shown that the delayed-rectifier Kv1.1 channel can be converted to A-type upon association with Kvbeta1.1 subunits; the conversion is only partial and is modulated by phosphorylation and microfilaments. Here we show that, in Xenopus oocytes, expression of Gbeta1gamma2 subunits concomitantly with the channel (composed of Kv1.1 and Kvbeta1.1 subunits), but not after the channel's expression in the plasma membrane, increases the extent of conversion to A-type. Conversely, scavenging endogenous Gbetagamma by co-expression of the C-terminal fragment of the beta-adrenergic receptor kinase reduces the extent of conversion to A-type. The effect of Gbetagamma co-expression is occluded by treatment with dihydrocytochalasin B, a microfilament-disrupting agent shown previously by us to enhance the extent of conversion to A-type, and by overexpression of Kvbeta1.1. Gbeta1gamma2 subunits interact directly with GST fusion fragments of Kv1.1 and Kvbeta1.1. Co-expression of Gbeta1gamma2 causes co-immunoprecipitation with Kv1.1 of more Kvbeta1.1 subunits. Thus, we suggest that Gbeta1gamma2 directly affects the interaction between Kv1.1 and Kvbeta1.1 during channel assembly which, in turn, disrupts the ability of the channel to interact with microfilaments, resulting in an increased extent of A-type conversion.