MilliporeSigma
  • Determination of HDL-cholesterol using 2,4,6-tribromo-3-hydroxybenzoic acid with a commercial CHOD-PAP reagent.
Determination of HDL-cholesterol using 2,4,6-tribromo-3-hydroxybenzoic acid with a commercial CHOD-PAP reagent.
Annals of clinical biochemistry (1984-09-01)
P Trinder, D Webster
ABSTRACT
The Boehringer one-component high-performance cholesterol oxidase reagent has been modified by the inclusion of 2,4,6-tribromo-3-hydroxybenzoic acid (TBHBA) to give a fourfold increase in sensitivity to a molar absorbance of approximately equal to 29,000 with respect to cholesterol. The resulting reagent system is particularly suitable for the determination of plasma HDL-cholesterol for which a reagent of high sensitivity is required. A simple method of bromination avoiding the use of elemental bromine is used to prepare TBHBA. The modified reagent system has been found to have good within- and between-batch precision and has shown itself to be reliable and trouble-free.
Sigma-Aldrich
2,4,6-Tribromo-3-hydroxybenzoic acid, 97%