MilliporeSigma
  • Graphene oxide-dispersed pristine CNTs support for MnO2 nanorods as high performance supercapacitor electrodes.
Graphene oxide-dispersed pristine CNTs support for MnO2 nanorods as high performance supercapacitor electrodes.
ChemSusChem (2013-02-19)
Bo You, Na Li, Hongying Zhu, Xiaolan Zhu, Jun Yang
ABSTRACT
A MnO2 -CNT-graphene oxide (MCGO) nanocomposite is fabricated using graphene oxide (GO) as a surfactant to directly disperse pristine carbon nanotubes (CNTs) for the subsequent deposition of MnO2 nanorods. The resulting MCGO nanocomposite is used as a supercapacitor electrode that shows ideal capacitive behavior (i.e., rectangular-shaped cyclic voltammograms), large specific capacitance (4.7 times higher than that of free MnO2 ) even at high mass loading (3.0 mg cm(-2) ), high energy density (30.4-14.2 Wh kg(-1) ), large power density (2.6-50.5 kW kg(-1) ), and still retains approximately 94 % of the initial specific capacitance after 1000 cycles. The advanced capacity, rate capability, and cycling stability may be attributed to the unique architecture, excellent ion wettability of GO with enriched oxygen-containing functional groups, high conductivity of CNTs, and their synergistic effects when combined with the other components. The results suggest that the MnO2 -CNT-GO hybrid nanocomposite architecture is very promising for next generation high-performance energy storage devices.
Sigma-Aldrich
Manganese(II,III) oxide, 97%