MilliporeSigma
  • Bicarbonate-dependent chloride transport drives fluid secretion by the human airway epithelial cell line Calu-3.
Bicarbonate-dependent chloride transport drives fluid secretion by the human airway epithelial cell line Calu-3.
The Journal of physiology (2012-07-11)
Jiajie Shan, Jie Liao, Junwei Huang, Renaud Robert, Melissa L Palmer, Scott C Fahrenkrug, Scott M O'Grady, John W Hanrahan
ABSTRACT
Anion and fluid secretion are both defective in cystic fibrosis (CF); however, the transport mechanisms are not well understood. In this study, Cl(-) and HCO(3)(-) secretion was measured using genetically matched CF transmembrane conductance regulator (CFTR)-deficient and CFTR-expressing cell lines derived from the human airway epithelial cell line Calu-3. Forskolin stimulated the short-circuit current (I(sc)) across voltage-clamped monolayers, and also increased the equivalent short-circuit current (I(eq)) calculated under open-circuit conditions. I(sc) was equivalent to the HCO(3)(-) net flux measured using the pH-stat technique, whereas I(eq) was the sum of the Cl(-) and HCO(3)(-) net fluxes. I(eq) and HCO(3)(-) fluxes were increased by bafilomycin and ZnCl(2), suggesting that some secreted HCO(3)(-) is neutralized by parallel electrogenic H(+) secretion. I(eq) and fluid secretion were dependent on the presence of both Na(+) and HCO(3)(-). The carbonic anhydrase inhibitor acetazolamide abolished forskolin stimulation of I(eq) and HCO(3)(-) secretion, suggesting that HCO(3)(-) transport under these conditions requires catalysed synthesis of carbonic acid. Cl(-) was the predominant anion in secretions under all conditions studied and thus drives most of the fluid transport. Nevertheless, 50-70% of Cl(-) and fluid transport was bumetanide-insensitive, suggesting basolateral Cl(-) loading by a sodium-potassium-chloride cotransporter 1 (NKCC1)-independent mechanism. Imposing a transepithelial HCO(3)(-) gradient across basolaterally permeabilized Calu-3 cells sustained a forskolin-stimulated current, which was sensitive to CFTR inhibitors and drastically reduced in CFTR-deficient cells. Net HCO(3)(-) secretion was increased by bilateral Cl(-) removal and therefore did not require apical Cl(-)/HCO(3)(-) exchange. The results suggest a model in which most HCO(3)(-) is recycled basolaterally by exchange with Cl(-), and the resulting HCO(3)(-)-dependent Cl(-) transport provides an osmotic driving force for fluid secretion.
Sigma-Aldrich
Bumetanide, ≥98%
Pricing and availability is not currently available.