MilliporeSigma
[Catheter-related bladder discomfort in post-anaesthesia care unit].
Annales francaises d'anesthesie et de reanimation (2012-07-04)
P Tauzin-Fin, L Stecken, F Sztark
ABSTRACT
Catheter-related bladder discomfort (CRBD) is an unrecognized clinical event. Symptoms of CRBD secondary to an indwelling urinary catheter mimic those of an overactive bladder, i.e. urinary frequency and urgency with or without urge incontinence. Stimulation of muscarinic receptors located in the bladder wall by the catheter is the triggering factor. Postoperative pain may be increased by the CRBD. Antimuscarinic drugs, as oxybutynin, are today the main treatment. Further studies are warranted to confirm efficacy of ketamine, tramadol and gabapentin in this situation.
Sigma-Aldrich
Oxybutynin chloride, meets EP, USP testing specifications
Sigma-Aldrich
Oxybutynin chloride, ≥98% (TLC), powder