MilliporeSigma
  • Hepato-protective effects of loganin, iridoid glycoside from Corni Fructus, against hyperglycemia-activated signaling pathway in liver of type 2 diabetic db/db mice.
Hepato-protective effects of loganin, iridoid glycoside from Corni Fructus, against hyperglycemia-activated signaling pathway in liver of type 2 diabetic db/db mice.
Toxicology (2011-08-26)
Chan Hum Park, Takashi Tanaka, Ji Hyun Kim, Eun Ju Cho, Jong Cheol Park, Naotoshi Shibahara, Takako Yokozawa
ABSTRACT
Accumulating evidence indicates that uncontrolled diabetes leads to the progression of diabetic complications such as liver disorder. The present study was carried out to elucidate the protective role of loganin extracted from Corni Fructus against hepatic oxidative stress caused by type 2 diabetes. Loganin (20 or 100mg/kg body weight/day, p.o.) was administered every day for 8 weeks to db/db mice, and its effect was assessed on comparison with vehicle-treated db/db and m/m mice. The administration of loganin led to a decrease in glucose and elevation of leptin in serum. The diabetic oxidative stress was attenuated by loganin through inhibitions of reactive oxygen species production and lipid peroxidation in the serum and liver. The expression of proteins induced by oxidative stress was significantly up-regulated in the liver of diabetic db/db mice; however, the expressions of both Nox-4 and p22(phox) were decreased significantly by loganin administration. Loganin showed a crucial effect in the inflammation-activated signaling pathway through the regulation of NF-κB, COX-2, and iNOS. It was also found to regulate the anti-inflammatory factors Nrf-2 and HO-1 in hepatic tissue. Moreover, expression of MCP-1 was significantly down-regulated in the loganin-treated db/db mice. Furthermore, loganin administration showed a protective effect against apoptosis by the regulation of Bcl-2 and cytochrome c. The present study demonstrated that the administration of loganin isolated from Corni Fructus had a protective effect against hepatic oxidative stress under type 2 diabetes through regulations of protein expressions related to oxidative stress, inflammation, and apoptosis.
Product Description
Supelco
Loganin, analytical standard
Pricing and availability is not currently available.