MilliporeSigma
  • Behavioural and insecticidal effects of organophosphate-, carbamate- and pyrethroid-treated mosquito nets against African malaria vectors.
Behavioural and insecticidal effects of organophosphate-, carbamate- and pyrethroid-treated mosquito nets against African malaria vectors.
Medical and veterinary entomology (2009-11-28)
R C Malima, R M Oxborough, P K Tungu, C Maxwell, I Lyimo, V Mwingira, F W Mosha, J Matowo, S M Magesa, M W Rowland
ABSTRACT
Three insecticides - the pyrethroid deltamethrin, the carbamate carbosulfan and the organophosphate chlorpyrifos-methyl - were tested on mosquito nets in experimental huts to determine their potential for introduction as malaria control measures. Their behavioural effects and efficacy were examined in Anopheles gambiae Giles s.s. (Diptera: Culicidae) and Anopheles funestus Giles s.s. in Muheza, Tanzania, and in Anopheles arabiensis Patton and Culex quinquefasciatus Say in Moshi, Tanzania. A standardized dosage of 25 mg/m(2) plus high dosages of carbosulfan (50 mg/m(2), 100 mg/m(2) and 200 mg/m(2)) and chlorpyrifos-methyl (100 mg/m(2)) were used to compare the three types of insecticide. At 25 mg/m(2), the rank order of the insecticides for insecticide-induced mortality in wild An. gambiae and An. funestus was, respectively, carbosulfan (88%, 86%) > deltamethrin (79%, 78%) > chlorpyrifos-methyl (35%, 53%). The rank order of the insecticides for blood-feeding inhibition (reduction in the number of blood-fed mosquitoes compared with control) in wild An. gambiae and An. funestus was deltamethrin > chlorpyrifos-methyl > carbosulfan. Carbosulfan was particularly toxic to endophilic anophelines at 200 mg/m(2), killing 100% of An. gambiae and 98% of An. funestus that entered the huts. It was less effective against the more exophilic An. arabiensis (67% mortality) and carbamate-resistant Cx quinquefasciatus (36% mortality). Carbosulfan deterred anophelines from entering huts, but did not deter carbamate-resistant Cx quinquefasciatus. Deltamethrin reduced the proportion of insects engaged in blood-feeding, probably as a consequence of contact irritancy, whereas carbosulfan seemed to provide personal protection through deterred entry or perhaps a spatial repellent action. Any deployment of carbosulfan as an individual treatment on nets should be carried out on a large scale to reduce the risk of diverting mosquitoes to unprotected individuals. Chlorpyrifos-methyl was inferior to deltamethrin in terms of mortality and blood-feeding inhibition and would be better deployed on a net in combination with a pyrethroid to control insecticide-resistant mosquitoes.
Product Description
Supelco
Chlorpyrifos-methyl, PESTANAL®, analytical standard