MilliporeSigma
Benzo(h)quinoline derivatives as G-quadruplex binding agents.
Bioorganic & medicinal chemistry letters (2009-02-27)
Hanumantharao Paritala, Steven M Firestine
ABSTRACT
G-quadruplexes are unusual structures formed from guanine-rich sequences of nucleic acids. G-quadruplexes have been postulated to play important roles in a number of biological systems including gene regulation and the inhibition of enzyme function. Recently, our laboratory reported on the synthesis and evaluation of a triaza-cyclopentaphenanthrene compound which bound to G-quadruplexes with good affinity and selectivity. This compound contains a 4-pyridone group which has not been previously utilized in other quadruplex binding agents. In this Letter, we describe the synthesis and evaluation of 4-pyridone containing 2- and 3-carboxy-benzoquinolines as G-quadruplex binding agents. We find that these compounds are capable of binding G-quadruplexes with a K(a) in the range of 3 x 10(5)M(-1) and with a 10-fold selectivity for quadruplex over duplex DNA.
Sigma-Aldrich
Benzo[h]quinoline, 97%
Pricing and availability is not currently available.