MilliporeSigma
Metabolism of glucurolactone to glucarate in Parkinson's disease.
Neurology (1992-09-01)
W C Koller, S D Rope, S J Huber, D C Blumenthal, H J Blumenthal
ABSTRACT
The metabolism of oral glucurolactone to serum D-glucarate by the hepatic cytochrome P450 enzyme system was no different in 20 untreated Parkinson's disease (PD) patients as compared with 20 age- and sex-matched controls. There was no evidence for a deficit in hepatic enzymes in PD.
Sigma-Aldrich
D-(+)-Glucuronic acid γ-lactone, ≥99%