MilliporeSigma
A study of peroxyoxalate-chemiluminescence of acriflavine.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2003-01-14)
Mojtaba Shamsipur, Mohammad Javad Chaichi, Ali Reza Karami
ABSTRACT
The chemiluminescence arising from the reaction of bis(2,4,6-trichlorophenyl)oxalate (TCPO) with hydrogen peroxide in the presence of acriflavine has been studied. The relationship between the chemiluminescence intensity and concentrations of TCPO, H2O2, acriflavine and the base sodium salicylate are reported. The kinetic parameters for the peroxyoxalate-chemiluminescence (PO-CL) of acriflavine were evaluated from the computer fitting of the corresponding chemiluminescence intensity-time plots.
Sigma-Aldrich
Bis(2,4,6-trichlorophenyl) oxalate, BioReagent, suitable for chemiluminescence, ≥99.0% (AT)