MilliporeSigma
Biomonitoring of exposure to lewisite based on adducts to haemoglobin.
Archives of toxicology (2000-08-26)
A Fidder, D Noort, A G Hulst, L P de Jong, H P Benschop
ABSTRACT
The development of a procedure for retrospective detection and quantitation of exposure to the arsenical dichloro(2-chlorovinyl)arsine (lewisite; L1) has been initiated. Upon incubation of human blood with [14C]L1 (20 nM-0.2 mM) in vitro, more than 90% of the total radioactivity was found in the erythrocytes and 25-50% of the radioactivity becomes associated with globin. Evidence was obtained for the presence of several binding sites. One type of binding was identified as L1-induced crosslinking of cysteine residues 93 and 112 of the beta-globin chain. A method was developed for extraction of bound and unbound 2-chlorovinylarsonous acid (CVAA), a major metabolite of L1, from whole blood after treatment with 2,3-dimercapto-1-propanol (BAL). Subsequent to derivatization with heptafluorobutyryl imidazole, the CVAA-BAL derivative could be analysed at a 40-fmol level by means of gas chromatography-mass spectroscopy (GC-MS) under electron impact conditions. With this procedure, in vitro exposure of human blood to 1 nM L1 could be determined. The same procedure was applied to the analysis of human urine samples spiked with CVAA. In vivo exposure of guinea pigs could be established at least 240 h after subcutaneous administration of the agent (0.25 mg/kg) by the determination of bound and unbound CVAA in the blood. In the urine of these animals, CVAA could be detected for 12 h after exposure.
Sigma-Aldrich
N-Heptafluorobutyrylimidazole, 97%
Sigma-Aldrich
1-(Heptafluorobutyryl)imidazole, BioReagent, suitable for derivatization