MilliporeSigma
  • Soluble epoxide hydrolase inhibitor, AUDA, prevents early salt-sensitive hypertension.
Soluble epoxide hydrolase inhibitor, AUDA, prevents early salt-sensitive hypertension.
Frontiers in bioscience : a journal and virtual library (2008-05-30)
Jing Li, Mairead A Carroll, Praveen N Chander, John R Falck, Bhavani Sangras, Charles T Stier
ABSTRACT
In stroke-prone spontaneously hypertensive rats (SHRSP) end-organ damage is markedly accelerated by high-salt (HS) intake. Since epoxyeicosatrienoic acids (EETs) possess vasodepressor and natriuretic activities, we examined whether a soluble epoxide hydrolase (sEH) inhibitor, 12-(3-adamantan-1-yl-ureido)-dodecanoic acid (AUDA), to inhibit the metabolism of EETs, would protect against pathologic changes in SHRSP. Seven-week-old male SHRSP were treated as follows: normal salt (NS), NS + AUDA, HS and HS + AUDA. Systolic blood pressure (SBP) (205 +/- 4 v 187 +/- 7 mmHg) and proteinuria (3.7 +/- 0.2 v 2.6 +/- 0.2 mg/6 h), but not plasma EETs (11.0 +/- 0.9 v 9.7 +/- 1.1 ng/ml), were significantly increased at 9 weeks of age in HS v NS SHRSP. HS was associated with fibrinoid degeneration and hypertrophy of arterioles in the kidney and perivascular fibrosis and contraction band necrosis in the heart. AUDA ameliorated these early salt-dependent changes in saline-drinking SHRSP and increased plasma levels of EETs but did not affect water and electrolyte excretion. sEH inhibition may provide a therapeutic strategy for treating salt-sensitive hypertension and its sequelae.
Sigma-Aldrich
AUDA, ≥98% (HPLC)
Pricing and availability is not currently available.