MilliporeSigma
  • 2-Chloro-1,4-dimethoxybenzene cation radical: formation and role in the lignin peroxidase oxidation of anisyl alcohol.
2-Chloro-1,4-dimethoxybenzene cation radical: formation and role in the lignin peroxidase oxidation of anisyl alcohol.
Archives of biochemistry and biophysics (1998-12-16)
P J Teunissen, D Sheng, G V Reddy, P Moënne-Loccoz, J A Field, M H Gold
ABSTRACT
2-Chloro-1,4-dimethoxybenzene (2Cl-1,4-DMB) oxidation by lignin peroxidase (LiP) proceeds via the formation of the 2Cl-1,4-DMB cation radical as indicated by ESR and UV/vis spectroscopy. The products of the LiP-catalyzed oxidation of 2Cl-1,4-DMB were identified as 2-chloro-1,4-benzoquinone and the dimers dichlorotetramethoxybiphenyl and chloro(chlorodimethoxyphenyl)benzoquinone. The addition of anisyl alcohol (AA) rapidly quenched the 2Cl-1,4-DMB cation radical optical absorption bands, suggesting that the cation radical directly mediates the oxidation of AA. When LiP reactions are conducted in the presence of 50 microM 2Cl-1,4-DMB, the enzyme is inactivated; however, this inactivation can be prevented by the addition of AA. This also suggests that the 2Cl-1,4-DMB cation radical formed in the reaction, in turn, oxidizes AA.
Sigma-Aldrich
2-Chloro-1,4-benzoquinone, 95%
Pricing and availability is not currently available.