MilliporeSigma
  • Cardiac anticholinergic effects of procainamide and its N-acetylated metabolite: experimental pharmacological and radioligand binding studies.
Cardiac anticholinergic effects of procainamide and its N-acetylated metabolite: experimental pharmacological and radioligand binding studies.
Journal of autonomic pharmacology (1998-09-08)
M Boucher, C Chassaing, P Laborde, C Breysse, A Herbet
ABSTRACT
1. The cardiac anticholinergic effects of procainamide (1 mg kg(-1) min(-1)) and its N-acetylated metabolite (NAPA) at equimolar dose (1.16 mg kg(-1) min(-1)) were studied using in vivo experimental pharmacological and in vitro radioligand binding studies. 2. Procainamide and NAPA progressively reduced vagal stimulation-induced bradycardia in chloralose-anaesthetized dogs. As indicated by the ED50, the vagolytic activity of NAPA is 1.5-2.0 times weaker than that of procainamide. Both drugs increased heart rate, with lowering of mean blood pressure during the second part of procainamide infusion, but not during NAPA infusion. 3. Binding studies on rat heart membranes yielded Ki values that were 1.5 times higher for NAPA than for procainamide. 4. These results show that NAPA exerts a weaker cardiac vagolytic action than procainamide, which is probably linked to a lower ability to bind to cardiac muscarinic receptors.
Sigma-Aldrich
N-Acetylprocainamide, ≥99%
Sigma-Aldrich
N-Acetylprocainamide hydrochloride, ≥99% (HPLC), powder