MilliporeSigma
  • Value of blood pool contrast agents in magnetic resonance angiography of the pelvis and lower extremities.
Value of blood pool contrast agents in magnetic resonance angiography of the pelvis and lower extremities.
European radiology (1998-07-31)
M Saeed, M F Wendland, M Engelbrecht, H Sakuma, C B Higgins
ABSTRACT
Our objective was: (1) to determine the appropriate dose of new ultrasmall superparamagnetic iron oxide particles for magnetic resonance angiography (MRA). This agent comprised of a single iron oxide crystal stabilized with a carbohydrate-polyethylene glycol coat (PEG-Ferron/NC 100150 injection); (2) to determine the proper flip angle for PEG-Ferron-enhanced 3 D time-of-flight (TOF) MRA sequence; and (3) to compare the enhancement of peripheral vessels following PEG-Ferron and GdDTPA-BMA. MRA parameters were: TR/TE = 50/2.1 ms, NEX = 1, FOV = 30 x 30 x 1.8 cm, and matrix = 256 x 128 x 64. In anesthetized beagle dogs (n = 10), the effects of PEG-Ferron and GdDTPA-BMA on regional signal were monitored for 45 min and compared. The lowest dose of PEG-Ferron (0.05 mmol/kg) produced the best enhancement of primary, secondary and tertiary vessels. The flip angle 60 degrees provided better enhancement than 20 degrees on contrast enhanced images. Unlike GdDTPA-BMA, PEG-Ferron allowed prolonged delineation (> 45 min) of the pelvis and lower extremities circulation. PEG-Ferron provided greater Contrast-to-noise ratio CNR (80.2 +/- 6.2, P < 0.05) than GdDTPA-BMA (63.5 +/- 2.5). It may be possible for blood pool contrast-enhanced 3 D TOF MRA to provide valuable information for visualization of vascular tree including guiding interventions.
Product Description
Supelco
8-Hydroxy-7-iodo-5-quinolinesulfonic acid, for spectrophotometric det. of Fe(III), ≥98.5%