MilliporeSigma
  • Synthesis of acylated methyl beta-D-xylopyranosides and their enzymic deacylations by rabbit serum esterases.
Synthesis of acylated methyl beta-D-xylopyranosides and their enzymic deacylations by rabbit serum esterases.
Carbohydrate research (1997-07-11)
V Petrović, S Tomić, D Ljevaković, J Tomasić
ABSTRACT
Selective pivaloylations of methyl beta-D-xylopyranoside have been studied under various reaction conditions. Partially pivaloylated products were submitted to additional acetylations. The structures were established by 1H NMR spectroscopy. Representatives of acylated methyl beta-D-xylopyranosides (acyl being pivaloyl, acetyl, or a combination of both) were submitted to hydrolysis catalyzed by rabbit serum and esterases isolated from rabbit serum.
Sigma-Aldrich
Methyl β-D-xylopyranoside, ≥99% (GC)