MilliporeSigma
  • Effects of low dose mixtures of four N-nitroso compounds on hepatic foci development in the rat.
Effects of low dose mixtures of four N-nitroso compounds on hepatic foci development in the rat.
Cancer letters (1996-09-10)
M Futakuchi, W Lijinsky, R Hasegawa, M Hirose, N Ito, T Shirai
ABSTRACT
Potential synergism between four N-nitroso compounds (nitrosomorpholine, nitrosodimethylamine, nitrosodiethanolamine, nitroso-oxazolidine) in rat liver carcinogenesis was examined in the medium-term bioassay. Male F344 rats were initially given diethylnitrosamine (DEN, 200 mg/kg, ip) and beginning 2 weeks later received test chemicals for 6 weeks individually at a full or 1/4 dose of that proven to be carcinogenic individually or in combination. All animals were subjected to partial hepatectomy at week 3 and killed at week 8. Induction of immunohistochemically-demonstrated glutathione S-transferase placental form (GST-P) positive foci was evaluated. The numbers and size of GST-P positive foci were significantly higher than the control levels by the treatment with each nitrosamine at full (1/1) and one quarter doses (1/4), excepting nitrosodiethanolamine and by combination of the four chemicals at 1/4 and 1/16. Because the dose-response curves were considered non-linear for most nitrosamines, synergistic effects were not apparent for the 1/4 mixture. Interestingly, however, the values for rats treated with these four chemicals in combination at the 1/4 dose level were almost the same as the average of four individual treatments at the full dose, and those for the 1/16 dose mixture were almost the same as the average of 1/4 individual treatment groups. These results indicate that these nitrosamines worked additively, rather than synergistically, in rat liver carcinogenesis.
Sigma-Aldrich
N-Nitrosodiethanolamine
Pricing and availability is not currently available.