MilliporeSigma
  • Reductive dehalogenation of the trichloromethyl group of nitrapyrin by the ammonia-oxidizing bacterium Nitrosomonas europaea.
Reductive dehalogenation of the trichloromethyl group of nitrapyrin by the ammonia-oxidizing bacterium Nitrosomonas europaea.
Applied and environmental microbiology (1993-11-01)
T Vannelli, A B Hooper
ABSTRACT
Suspensions of Nitrosomonas europaea catalyzed the reductive dehalogenation of the commercial nitrification inhibitor nitrapyrin (2-chloro-6-trichloromethylpyridine). The product of the reaction was identified as 2-chloro-6-dichloromethylpyridine by its mass fragmentation and nuclear magnetic resonance spectra. A small amount of 2-chloro-6-dichloromethylpyridine accumulated during the conversion of nitrapyrin to 6-chloropicolinic acid in an aerated solution in the presence of ammonia (T. Vannelli and A.B. Hooper, Appl. Environ. Microbiol. 58:2321-2325, 1992). Nearly stoichiometric conversion of nitrapyrin to 2-chloro-6-dichloromethylpyridine occurred at very low oxygen concentrations and in the presence of hydrazine as a source of electrons. Under these conditions the turnover rate was 0.37 nmol of nitrapyrin per min per mg of protein. Two specific inhibitors of ammonia oxidation, acetylene and allylthiourea, inhibited the rate of the dehalogenation reaction by 80 and 84%, respectively. In the presence of D2O, all 2-chloro-6-dichloromethylpyridine produced in the reaction was deuterated at the methyl position. In an oxygenated solution and in the presence of ammonia or hydrazine, cells did not catalyze the oxidation of exogenously added 2-chloro-6-dichloromethylpyridine to 6-chloropicolinic acid. Thus, 2-chloro-6-dichloromethylpyridine is apparently not an intermediate in the aerobic production of 6-chloropicolinic acid from nitrapyrin.
Product Description
Supelco
Nitrapyrin, PESTANAL®, analytical standard
Sigma-Aldrich
2-Chloro-6-(trichloromethyl)pyridine, ≥98%