MilliporeSigma
  • Spermidine acetylation in N1 and N8 position in rat brain and in N-ethyl-N-nitrosourea-induced gliomas.
Spermidine acetylation in N1 and N8 position in rat brain and in N-ethyl-N-nitrosourea-induced gliomas.
Cancer letters (1994-01-30)
A Sessa, E Broglia, M R Terreni, A Perin
ABSTRACT
N-ethyl-N-nitrosourea-induced rat gliomas showed a stimulation of cytosolic spermidine N-acetyltransferase activity compared with normal brain, with an increased formation of N1 and N8-acetylspermidine, suggesting the activation of two enzymes acetylating the polyamine in N1 and N8 position, respectively. The enhancement in cytosolic spermidine N1-acetyltransferase was probably responsible for an activation of the polyamine interconversion pathway in gliomas, as indicated by an accumulation of N1-acetylspermidine and a marked increase in putrescine. Spermidine N8-acetyltransferase, although acetylating histones at a high rate, seemed distinct from the nuclear enzyme, as, unlike from the latter, it showed very low affinity for putrescine and was not inhibited by methylglyoxal bis(guanylhydrazone).
Product Description
Supelco
N8-Acetylspermidine dihydrochloride, analytical standard