MilliporeSigma
  • Electrophysiological evidence for a functional interaction between 5-HT1A and 5-HT2A receptors in the rat medial prefrontal cortex: an iontophoretic study.
Electrophysiological evidence for a functional interaction between 5-HT1A and 5-HT2A receptors in the rat medial prefrontal cortex: an iontophoretic study.
Synapse (New York, N.Y.) (1994-07-01)
C R Ashby, E Edwards, R Y Wang
ABSTRACT
In this study, we examined the interaction of 5-HT1A and 5-HT2A receptors in the rat medial prefrontal cortex (mPFc) using the techniques of extracellular single unit recording and microiontophoresis. The iontophoresis of the selective 5-HT1A receptor agonist (+-)-8-hydroxy-2-(di-n-propylamino) tetralin (8-OHDPAT) produced a current-dependent suppression (2.5-20 nA) of the basal firing rate of spontaneously active mPFc cells. The iontophoretic (5-10 nA) and systemic administration (0.1-0.5 mg/kg, i.v.) of the 5-HT2A/5-HT2C receptor antagonist ritanserin and the selective 5-HT2A receptor antagonist MDL 28727 significantly potentiated and prolonged 8-OHDPAT's suppressant action. In addition, the systemic administration of another selective 5-HT2A antagonist MDL 100907, but not its less active enantiomer MDL 100009, also potentiated and prolonged 8-OHDPAT's action. The potentiating effect of the 5-HT2A receptor antagonists on the action of 8-OHDPAT is specific in that neither the iontophoresis of ritanserin nor MDL 28727 altered the suppressant action produced by the iontophoresis of the 5-HT3 receptor agonist 2-methylserotonin onto mPFc cells. Moreover, the suppressant action of 8-OHDPAT was not altered by the systemic administration of the selective 5-HT3 receptor antagonist granisetron (0.1-0.5 mg/kg, i.v.). On the other hand, the iontophoresis of a low current (0.5 nA) of the 5-HT2A,2C receptor agonist (+-)-1-(2,5-dimethoxy-4-iodophenyl)-2-aminopropane (DOI) potentiated the excitation induced by the iontophoresis of l-glutamate on quiescent mPFc cells.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
5-Hydroxy--methyltryptamine oxalate, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Nω-Methyl-5-hydroxy­trypt­amine oxalate salt, ≥90% (HPLC)
Pricing and availability is not currently available.