MilliporeSigma
  • Inhibition of DMBA carcinogenesis of hamster buccal pouch by phenanthrene and dimethylnaphthalene.
Inhibition of DMBA carcinogenesis of hamster buccal pouch by phenanthrene and dimethylnaphthalene.
Carcinogenesis (1981-01-01)
D S Malament, G Shklar
ABSTRACT
Phenanthrene (Phe) and to a lesser degree 1,4-dimethylnaphthalene (DMeN) were each found to retard the development of epidermoid carcinomas in hamster buccal pouch induced by the thrice weekly application of a 0.5 per cent solution of 7,12-dimethylbenz[a]anthracene (DMBA) in heavy mineral oil.
Sigma-Aldrich
1,4-Dimethylnaphthalene, 95%
Pricing and availability is not currently available.