MilliporeSigma
  • Effect of ascorbic acid deficiency on the in vivo synthesis of carnitine.
Effect of ascorbic acid deficiency on the in vivo synthesis of carnitine.
Biochimica et biophysica acta (1981-01-07)
P J Nelson, R E Pruitt, L L Henderson, R Jenness, L M Henderson
ABSTRACT
The effects of ascorbate deficiency on carnitine biosynthesis was investigated in young male guinea pigs. Liver and kidney carnitine levels were not affected by the deficiency, but scorbutic animals had 50% less carnitine in heart and skeletal muscle than control animals. Labeled carnitine precursors, 6-N-tri-methyl-L-lysine and 4-N-trimethylaminobutyrate, both of which require ascorbate for their enzymatic hydroxylation, were injected into the vena cava of control, pair-fed and scorbutic animals. The distribution of isotope in compounds present in the liver and kidney after 1 h was determined. The uptake of trimethyllysine by the liver was less than 2% in 1 h, while the kidney took up approx. 20% of the 14C. Control and pair-fed animals converted trimethyllysine to kidney trimethylaminobutyrate 8--10 times as well as did scorbutic animals. Trimethylaminobutyrate hydroxylase, present in the liver but almost absent from the kidney, converted nearly all of substrate taken up by the liver to carnitine in both the scorbutic and control animals.
Sigma-Aldrich
(3-Carboxypropyl)trimethylammonium chloride, technical grade
Pricing and availability is not currently available.