MilliporeSigma
  • Antitumor activity of bis(diphenylphosphino)alkanes, their gold(I) coordination complexes, and related compounds.
Antitumor activity of bis(diphenylphosphino)alkanes, their gold(I) coordination complexes, and related compounds.
Journal of medicinal chemistry (1987-12-01)
C K Mirabelli, D T Hill, L F Faucette, F L McCabe, G R Girard, D B Bryan, B M Sutton, J O Bartus, S T Crooke, R K Johnson
ABSTRACT
Bisphosphines related to bis(diphenylphosphino)ethane (dppe) and their gold complexes are described that are active in a spectrum of transplantable tumor models. When administered ip on days 1-5 at its maximally tolerated dose (MTD) of 40 mumol/kg, dppe reproducibly gives 100% increase in life span (ILS) in mice bearing ip P388 leukemia. Coordination of chlorogold(I) to each phosphine in dppe gave a complex that had similar activity but at a much lower dose level than dppe; the MTD for the gold(I) complex was 7 mumol/kg. Among other metal complexes of dppe, the Au(III) complex was active (greater than 50% ILS) whereas Ag(I), Ni(II), Pt(II), Pd(II), and Rh(I) complexes were inactive. Among dppe analogues, replacement of phenyl groups with ethyl or benzyl groups resulted in inactivity for both ligands and the corresponding gold complexes whereas substitution with cyclohexyl or heterocyclic ring systems yielded ligands and/or gold complexes with antitumor activity. Among substituted-phenyl dppe and dppe(AuCl)2 analogues, 3-fluoro, 4-fluoro, perdeuterio, 4-methylthio, and 2-methylthio analogues were active; 4-methyl, 3-methyl, 4-methoxy, 4-dimethylamino, and 4-trifluoromethyl analogues were marginal or inactive. Analogues in which the ethane bridge of dppe or dppe(AuCl)2 was varied between one and six carbons, unsaturated or substituted, revealed that activity was maximal with ethane or cis-ethylene. Compounds with good P388 activity were also active in other animal tumor models.
Sigma-Aldrich
Ethylenebis(diphenylphosphine), 99%
Sigma-Aldrich
Ethylenebis(diphenylphosphine), 97%