MilliporeSigma
  • Analyses of azopigments obtained from the delta fraction of bilirubin from mammalian plasma (mammalian biliprotein).
Analyses of azopigments obtained from the delta fraction of bilirubin from mammalian plasma (mammalian biliprotein).
The Biochemical journal (1987-11-15)
H Yoshida, T Inagaki, M Hirano, T Sugimoto
ABSTRACT
Azopigments were obtained from the delta fraction of bilirubin (mammalian biliprotein) in cholestatic sera of men, rats and guinea pigs by diazo reaction with diazotized p-iodoaniline and analysed by t.l.c. Delta bilirubin of men and rats generated both unconjugated and glucuronide-conjugated azodipyrroles, whereas that of guinea pigs, in which the predominant form of conjugated bilirubin in serum was bilirubin monoglucuronide, generated only unconjugated azodipyrrole. We further analysed the azopigments by reversed-phase h.p.l.c. to distinguish their endovinyl and exovinyl isomers. The results indicated (a) that covalent binding of bilirubin to protein occurs exclusively on the conjugated dipyrrolic (either endovinyl or exovinyl) half of the parent conjugated bilirubin, (b) that both bilirubin monoglucuronide and bilirubin diglucuronide generate delta bilirubin, the latter yielding a 'conjugated' form of delta bilirubin that preserves the glucuronic acid moiety on the dipyrrolic half not bound covalently to protein, and (c) that therefore at least four forms of delta bilirubin exist in jaundiced sera of men and rats.
Sigma-Aldrich
4-Iodoaniline, 98%
Pricing and availability is not currently available.