MilliporeSigma
  • The effect of temperature of fluorescence for liquid scintillators and their solvents.
The effect of temperature of fluorescence for liquid scintillators and their solvents.
International journal of radiation applications and instrumentation. Part A, Applied radiation and isotopes (1987-01-01)
Y Homma, Y Murase, K Sonehara
ABSTRACT
The pulse-height distributions for 241Am and 131mXe in PPO solutions of the aromatic hydrocarbons, i.e. benzene, o-xylene, m-xylene, ethylbenzene and cumene are found to shift toward higher pulse-heights with decreasing temperature. Under u.v. excitation, the fluorescence intensity of these pure aromatic hydrocarbons increases markedly and the fluorescence maximum is found to shift to longer wavelengths with decreasing temperature. In conjunction with observations of the pulse-height shift in liquid scintillators and the fluorescence emission from the pure solvents, the pulse-height shifts observed are interpreted in terms of excimer formation in the aromatic hydrocarbons.
Sigma-Aldrich
2,5-Diphenyloxazole, suitable for liquid scintillation spectrometry
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2,5-Diphenyloxazole, 99%, suitable for scintillation