MilliporeSigma
Fourier transform infrared assay of liposomal lipids.
Analytical biochemistry (1989-08-15)
C Pidgeon, G Apostol, R Markovich
ABSTRACT
Quantitating the lipid content in organic lipid solutions and extracted membrane preparations is described. Fourier transform infrared analysis of thin films using perdeuterated nonadecane as an internal standard permitted quantitation with greater than 95% accuracy.
Product Description
Supelco
Nonadecane, analytical standard
Sigma-Aldrich
Nonadecane, 99%