MilliporeSigma
  • Effect of ofurace, oxadixyl, and alachlor on xenobiotic biotransformation in the rat liver.
Effect of ofurace, oxadixyl, and alachlor on xenobiotic biotransformation in the rat liver.
Archives of environmental contamination and toxicology (1989-11-01)
C Leslie, G F Reidy, N H Stacey
ABSTRACT
Ofurace, oxadixyl, and alachlor were studied for their effect on hepatic xenobiotic biotransformation in male rats by dosing i.p. with 1 or 100 mg/kg of chemical for 7 days. Ofurace decreased ethoxyresorufin-O-deethylase and aniline p-hydroxylase activities but induced ethoxycoumarin-O-deethylase activity. Glutathione S-transferase and aminopyrine N-demethylase activities responded in a non dose-dependent manner, while cytochrome P-450 content and aldrin epoxidase activities were unchanged. Oxadixyl induced P-450 content, and ethoxycoumarin-O-deethylase and aminopyrine N-demethylase activities. It left ethoxyresorufin-O-deethylase, aldrin epoxidase and epoxide hydrolase activities unchanged and decreased aniline p-hydroxylase activities. Alachlor induced all activities excepting aldrin epoxidase (no change) and aniline p-hydroxylase (decreased). The data indicate that each of these acylanilide pesticides interacts with the monooxygenase system but with differing patterns.
Product Description
Supelco
Ofurace, PESTANAL®, analytical standard
Supelco
Oxadixyl, PESTANAL®, analytical standard