MilliporeSigma
  • Determination of thiols and disulfides in normal rat tissues and hamster pancreas treated with N-nitrosobis(2-oxopropyl)amine using 4-(aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole and ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate.
Determination of thiols and disulfides in normal rat tissues and hamster pancreas treated with N-nitrosobis(2-oxopropyl)amine using 4-(aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole and ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate.
Biomedical chromatography : BMC (1989-07-01)
T Toyo'oka, F Furukawa, T Suzuki, Y Saito, M Takahashi, Y Hayashi, S Uzu, K Imai
ABSTRACT
Biological thiols and disulfides in rat and hamster tissues were simultaneously determined by HPLC-fluorescence detection using 4-(aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole (ABD-F) and ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate (SBD-F). The coefficients of variation (CV) of the method for reduced glutathione (GSH) and oxidized glutathione (GSSG) in liver and for cysteine (CySH) and cystine (CySSCy) in kidney were less than 3.1%. In 11 tissues of Wistar rats (liver, spleen, heart, lung, stomach, bladder, ovary, uterus, adrenal, kidney and pancreas), only CySH, CySSCy, GSH and/or GSSG were detected. Other thiols and disulfides were at extremely low levels in all samples. Both concentrations of CySH and CySSCy in the livers of old rats (111 weeks old, F344) were significantly higher than those of young rats (8 weeks old) (CySH, 0.246 +/- 0.099 vs 0.130 +/- 0.020 mumol/g; CySSCy, 0.051 +/- 0.027 vs 0.013 +/- 0.002 mumol/g). Administration of N-nitrosobis(2-oxopropyl)amine (BOP), a selective carcinogen of hamster pancreatic cancer, to Syrian golden hamsters (38 weeks old) resulted in the increase in the pancreas of GSH to a level 19 times as high and of GSSG to a level 14 times as high as those in untreated hamsters (GSH, 1.173 +/- 0.272 vs 0.062 +/- 0.017 mumol/g; GSSG, 0.155 +/- 0.063 vs 0.011 +/- 0.001 mumol/g).
Sigma-Aldrich
7-Fluorobenzofurazan-4-sulfonic acid ammonium salt, ≥98%
Supelco
7-Fluorobenzofurazan-4-sulfonic acid ammonium salt, for HPLC derivatization, LiChropur, ≥98.5% (HPLC)