MilliporeSigma
  • Photocatalytic reduction of Cr(VI) by citric and oxalic acids over biogenetic jarosite.
Photocatalytic reduction of Cr(VI) by citric and oxalic acids over biogenetic jarosite.
Materials science & engineering. C, Materials for biological applications (2013-03-19)
Zhihui Xu, Shuangyou Bai, Jianru Liang, Lixiang Zhou, Yeqing Lan
ABSTRACT
In this study, a series of bath experiments were carried out to investigate the photoreduction of Cr(VI) by small molecular weight organic acids (SOAs) over jarosite, a mineral found in acid mine drainage (AMD). The results demonstrated that jarosite or SOAs alone was unable to effectively transform Cr(VI) to Cr(III) even if exposed to an illumination of mimic solar light. However, an addition of jarosite significantly enhanced the reduction of Cr(VI) by SOAs under the same condition. The photocatalytic reduction of Cr(VI) was strongly influenced by pH, the initial concentrations and the structures of SOAs. Of the tested two SOAs, the reaction rates of photocatalytic reduction of Cr(VI) were in the order of oxalic acid>citric acid. The reaction obeyed to zero-order kinetics with respect to Cr(VI) with excess SOAs. A possible mechanism for photoreduction of Cr(VI) by SOAs over jarosite was proposed.
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Sigma-Aldrich
Oxalic acid, SAJ first grade, ≥97.0%
Sigma-Aldrich
Oxalic acid solution, 0.05 M
Sigma-Aldrich
Oxalic acid solution, 0.5 M
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Supelco
Sodium oxalate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Sodium oxalate, ≥99.99% trace metals basis
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium oxalate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium oxalate, puriss. p.a., ACS reagent, ≥99.5% (RT)
Sigma-Aldrich
Sodium oxalate, BioXtra
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Sodium oxalate solution, 0.0125 M
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium oxalate solution, 5 mM
Sigma-Aldrich
Citric acid, SAJ first grade, ≥99.5%
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland