- Aqueous synthesis of N,S-dialkylthiophosphoramidates: design, optimisation and application to library construction and antileishmanial testing.
Aqueous synthesis of N,S-dialkylthiophosphoramidates: design, optimisation and application to library construction and antileishmanial testing.
Organic & biomolecular chemistry (2013-03-08)
Milena Trmčić, Frances L Chadbourne, Paul M Brear, Paul W Denny, Steven L Cobb, David R W Hodgson
PMID23467665
ABSTRACT
We recently reported the use of PSCl3 for the thiophosphorylation of alkylamines where the resulting N-thiophosphoramidate ions could be readily S-alkylated (Chem. Commun., 2011, 47, 6156-6158.). Herein we report the development of this methodology using amino acid, amino sugar, aminonucleoside and aniline substrates. The hydrolysis properties of N-thiophosphoramidate ions and their reactivities towards alkylating agents are also explored. In addition, we demonstrate the application of our approach to the preparation of a small library of compounds, including quinoline-based N,S-dialkylthiophosphoramidates which were tested for antileishmanial activity.
Product Description
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%