Sigma-Aldrich
Nitrilotriacetic acid, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Ethylene glycol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Pricing and availability is not currently available.
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol 5 M solution
Pricing and availability is not currently available.