MilliporeSigma
  • Apoptosis of human breast cancer cells induced by microencapsulated betulinic acid from sour jujube fruits through the mitochondria transduction pathway.
Apoptosis of human breast cancer cells induced by microencapsulated betulinic acid from sour jujube fruits through the mitochondria transduction pathway.
Food chemistry (2013-02-16)
Yan-Fang Sun, Chuan-Kui Song, Helmut Viernstein, Frank Unger, Zong-Suo Liang
ABSTRACT
Betulinic acid (BA), a natural pentacyclic triterpenoid, was isolated from sour jujube fruits for the first time. An inclusion complex comprising BA and β-cyclodextrin (β-CD) was formed to improve the dissolution of BA, but little is known about its anticancer effect. In this study, the anti-proliferative and apoptosis mechanisms of BA-β-CD on human breast cancer MCF-7 cells were further investigated. Experimental results confirmed that the complexation model inhibited the growth of MCF-7 cells in a dose-dependent manner, arrested cell cycle in the G2/M phase and induced apoptosis via the mitochondria transduction pathway. Gene and protein analyses showed that the complexation model significantly inhibited Bcl-2 expression and promoted Bax expression, causing caspase-3 and caspase-9 cascade activation. These findings corroborated evidence on microencapsulated BA as an apoptosis inducer in MCF-7 cells. Thus, sour jujube fruits may have potential use as a breast cancer chemotherapeutic agent.
Sigma-Aldrich
Betulinic acid, ≥98% (HPLC)
Supelco
Betulinic acid, analytical standard
Sigma-Aldrich
Betulinic acid, technical grade, 90%