MilliporeSigma
  • The N-terminal fragment of Acanthamoeba polyphaga mimivirus tyrosyl-tRNA synthetase (TyrRS(apm)) is a monomer in solution.
The N-terminal fragment of Acanthamoeba polyphaga mimivirus tyrosyl-tRNA synthetase (TyrRS(apm)) is a monomer in solution.
FEBS letters (2013-02-07)
Aparajita Choudhury, Rajat Banerjee
ABSTRACT
Acanthamoeba polyphaga mimivirus tyrosyl-tRNA synthetase (TyrRSapm) was the first reported aminoacyl-tRNA synthetase of viral origin. The previous crystal structure of TyrRSapm showed a non-canonical orientation of the dimer conformation and the CP1 domain, responsible for dimer formation, displays a major modification of a motif structurally conserved in other TyrRS structures. An earlier study reported that Bacillus stearothermophilus N-terminal TyrRS exists as a dimer under native conditions. N-terminal TyrRSapm (ΔTyrRSapm, 1-234 aa) was constructed to remove the C-terminal anticodon-binding domain. Here we show by Ferguson plot analysis and analytical ultracentrifugation that ΔTyrRSapm exists as a monomer and contains a disulfide-bridge. The ΔTyrRSapm loses the ability to bind tRNA(Tyr), however it remains active in pyrophosphate exchange with similar ligand dissociation constants as the full-length enzyme.
Sigma-Aldrich
Pyrophosphoric acid, SAJ first grade, ≥90.0%
Sigma-Aldrich
Pyrophosphoric acid, technical grade