MilliporeSigma
  • The characteristics and chronology of the earliest Acheulean at Konso, Ethiopia.
The characteristics and chronology of the earliest Acheulean at Konso, Ethiopia.
Proceedings of the National Academy of Sciences of the United States of America (2013-01-30)
Yonas Beyene, Shigehiro Katoh, Giday Woldegabriel, William K Hart, Kozo Uto, Masafumi Sudo, Megumi Kondo, Masayuki Hyodo, Paul R Renne, Gen Suwa, Berhane Asfaw
ABSTRACT
The Acheulean technological tradition, characterized by a large (>10 cm) flake-based component, represents a significant technological advance over the Oldowan. Although stone tool assemblages attributed to the Acheulean have been reported from as early as circa 1.6-1.75 Ma, the characteristics of these earliest occurrences and comparisons with later assemblages have not been reported in detail. Here, we provide a newly established chronometric calibration for the Acheulean assemblages of the Konso Formation, southern Ethiopia, which span the time period ∼1.75 to <1.0 Ma. The earliest Konso Acheulean is chronologically indistinguishable from the assemblage recently published as the world's earliest with an age of ∼1.75 Ma at Kokiselei, west of Lake Turkana, Kenya. This Konso assemblage is characterized by a combination of large picks and crude bifaces/unifaces made predominantly on large flake blanks. An increase in the number of flake scars was observed within the Konso Formation handaxe assemblages through time, but this was less so with picks. The Konso evidence suggests that both picks and handaxes were essential components of the Acheulean from its initial stages and that the two probably differed in function. The temporal refinement seen, especially in the handaxe forms at Konso, implies enhanced function through time, perhaps in processing carcasses with long and stable cutting edges. The documentation of the earliest Acheulean at ∼1.75 Ma in both northern Kenya and southern Ethiopia suggests that behavioral novelties were being established in a regional scale at that time, paralleling the emergence of Homo erectus-like hominid morphology.
Sigma-Aldrich
Argon, ≥99.998%