MilliporeSigma
CO oxidation on h-BN supported Au atom.
The Journal of chemical physics (2013-01-25)
Min Gao, Andrey Lyalin, Tetsuya Taketsugu
ABSTRACT
The mechanism of CO oxidation by O(2) on Au atoms supported on the pristine and defected hexagonal boron nitride (h-BN) surface has been studied theoretically using density functional theory. It is found that O(2) binds stronger than CO on an Au atom supported on the defect free h-BN surface and h-BN surface with nitrogen vacancy (V(N)@h-BN), but weaker than CO on a free Au atom or Au trapped by a boron vacancy (V(B)@h-BN). The excess of the positive or negative charge on Au can considerably change its catalytic properties and enhance activation of the adsorbed O(2). Coadsorption of CO and O(2) on Au, Au/V(N)@h-BN, and Au/V(B)@h-BN results in additional charge transfer to O(2). Various pathways of the CO oxidation reaction by molecular oxygen are studied. We found two different pathways for CO oxidation: a two-step pathway where two CO(2) molecules are formed independently, and a self-promotion pathway where oxidation of the first CO molecule is promoted by the second CO molecule. Interaction of Au with the defect-free and defected h-BN surface considerably affects the CO oxidation reaction pathways and barriers. Therefore, Au supported on the h-BN surface (pristine or defected) cannot be considered as pseudo-free atom and support effects have to be taken into account, even when the interaction of Au with the support is weak.
Sigma-Aldrich
Boron nitride, powder, ~1 μm, 98%