MilliporeSigma
  • Understanding self-photorechargeability of WO(3) for H(2) generation without light illumination.
Understanding self-photorechargeability of WO(3) for H(2) generation without light illumination.
ChemSusChem (2013-01-17)
Charlene Ng, Akihide Iwase, Yun Hau Ng, Rose Amal
ABSTRACT
This work presents insight into the self-photorechargeability of WO(3), whereby the intercalation of positive alkali cations is accompanied by the simultaneous storage of photo-excited electrons. The cyclic voltammetry studies verify the photo-assisted intercalation and de-intercalation of Na(+) and K(+) from the flower structured WO(3). A storage capacity of up to 0.722 C cm(-2) can be achieved in a saturated (0.68 M) K(2)SO(4) electrolyte solution. However, the best photo recharge-discharge stability of the electrode are observed at a lower (0.1 M) cation concentration. At high electrolyte concentrations, the intercalated cations are firmly trapped, as indicated by the structural modifications observed in Raman analysis, resulting in much less photocharging and discharging abilities in subsequent cycles. The study also shows that the stored electrons can be successfully used to generate H(2) with 100 % faradaic efficiency in the absence of light.
Sigma-Aldrich
Sodium sulfate, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium hydrogen sulfate, technical grade
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Tungsten, powder (monocrystalline), 0.6-1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Tungsten, powder, 12 μm, 99.9% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Potassium sulfate, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Tungsten, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate decahydrate, BioUltra, ≥99.0% (calc. on dry substance, T)
Sigma-Aldrich
Potassium sulfate, 99.99% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Potassium bisulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Tungsten, powder, ≤10 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium sulfate, suitable for plant cell culture
Sigma-Aldrich
Tungsten, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.127 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium sulfate decahydrate, puriss. p.a., crystallized, ≥99.0% (calc. based on dry substance, T)
Sigma-Aldrich
Potassium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium bisulfate monohydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Potassium sulfate, meets analytical specification of Ph. Eur., 99-101%