MilliporeSigma
  • Synthesis of a branched glycopeptide derivative containing terminal D-mannose 6-phosphate residues.
Synthesis of a branched glycopeptide derivative containing terminal D-mannose 6-phosphate residues.
Carbohydrate research (1990-05-01)
Y Ichikawa, Y C Lee
ABSTRACT
A branched glycopeptide derivative incorporating two D-mannose 6-phosphate residues was prepared by coupling 6-aminohexyl 6-O-[bis(2,2,2-trichloroethoxy)phosphinyl]-alpha-D-mannopyranoside with N-acetyl-L-tyrosyl-L-aspart-oyldi-L-alanine followed by reductive deprotection of the phosphate group.