MilliporeSigma
  • QLT091001, a 9-cis-retinal analog, is well-tolerated by retinas of mice with impaired visual cycles.
QLT091001, a 9-cis-retinal analog, is well-tolerated by retinas of mice with impaired visual cycles.
Investigative ophthalmology & visual science (2012-12-20)
Tadao Maeda, Zhiqian Dong, Hui Jin, Osamu Sawada, Songqi Gao, Deepank Utkhede, Wendy Monk, Grazyna Palczewska, Krzysztof Palczewski
ABSTRACT
Investigate whether retinas of mice with impaired retinal cycles exposed to light or kept in the dark tolerate prolonged high-dose administration of QLT091001, which contains as an active ingredient, the 9-cis-retinal precursor, 9-cis-retinyl acetate. Four- to six-week-old Lrat(-/-) and Rpe65(-/-) mice (n = 126) as well as crossbred Gnat1(-/-) mice lacking rod phototransduction (n = 110) were gavaged weekly for 6 months with 50 mg/kg QLT091001, either after being kept in the dark or after light bleaching for 30 min/wk followed by maintenance in a 12-hour light ≤ 10 lux)/12-hour dark cycle. Retinal health was monitored by spectral-domain optical coherent tomography (SD-OCT) and scanning laser ophthalmoscopy (SLO) every other month and histological, biochemical, and visual functional analyses were performed at the end of the experiment. Two-photon microscopy (TPM) was used to observe retinoid-containing retinosome structures in the RPE. Retinal thickness and morphology examined by SD-OCT were well maintained in all strains treated with QLT091001. No significant increases of fundus autofluorescence were detected by SLO imaging of any strain. Accumulation of all-trans-retinyl esters varied with genetic background, types of administered compounds and lighting conditions but retinal health was not compromised. TPM imaging clearly revealed maintenance of retinosomes in the RPE of all mouse strains tested. Retinas of Lrat(-/-), Rpe65(-/-), and crossbred Gnat1(-/-) mice tolerated prolonged high-dose QLT091001 treatment well.
Sigma-Aldrich
9-cis-Retinal, vitamin A analog
Sigma-Aldrich
all trans-Retinal, powder, ≥98%