MilliporeSigma
  • Wogonin inhibits H2O2-induced vascular permeability through suppressing the phosphorylation of caveolin-1.
Wogonin inhibits H2O2-induced vascular permeability through suppressing the phosphorylation of caveolin-1.
Toxicology (2012-12-19)
Fei Wang, Xiuming Song, Mi Zhou, Libin Wei, Qinsheng Dai, Zhiyu Li, Na Lu, Qinglong Guo
ABSTRACT
Wogonin, a naturally occurring monoflavonoid extracted from the root of Scutellaria baicalensis Georgi, has been reported for its anti-oxidant activity. However, it is still unclear whether wogonin can inhibit oxidant-induced vascular permeability. In this study, we evaluated the effects of wogonin on H2O2-induced vascular permeability in human umbilical vein endothelial cells (HUVECs). We found that wogonin can suppress the H2O2-stimulated actin remodeling and albumin uptake of HUVECs, as well as transendothelial cell migration of the human breast carcinoma cell MDA-MB-231. The mechanism revealed that wogonin inhibited H2O2-induced phosphorylation of caveolin-1 (cav-1) associating with the suppression of stabilization of VE-cadherin and β-catenin. Moreover, wogonin repressed anisomycin-induced phosphorylation of p38, cav-1 and vascular permeability. These results suggested that wogonin could inhibit H2O2-induced vascular permeability by downregulating the phosphorylation of cav-1, and that it might have a therapeutic potential for the diseases associated with the development of both oxidant and vascular permeability.
Sigma-Aldrich
Wogonin hydrate, ≥98% (HPLC), solid