- Study of algal biomass harvesting using cationic guar gum from the natural plant source as flocculant.
Study of algal biomass harvesting using cationic guar gum from the natural plant source as flocculant.
Carbohydrate polymers (2012-12-12)
Chiranjib Banerjee, Sandipta Ghosh, Gautam Sen, Sumit Mishra, Pratyoosh Shukla, Rajib Bandopadhyay
PMID23218353
ABSTRACT
Microalgae are small in size with negatively charged surface. They are usually stable in suspension culture and hard to flocculate. The present work emphasizes on the synthesis of cationic guar gum (CGG) by the introduction of quaternary amine groups onto the backbone of guar gum (GG) from N-(3-chloro-2-hydroxypropyl) trimethyl ammonium chloride (CHPTAC). The optimal dosage of the synthesized cationic guar gum is used to flocculate two different green algae viz. Chlorella sp. CB4 and Chlamydomonas sp. CRP7.
MATERIALS