MilliporeSigma
  • Mathematical modeling of triphasic viral dynamics in patients with HBeAg-positive chronic hepatitis B showing response to 24-week clevudine therapy.
Mathematical modeling of triphasic viral dynamics in patients with HBeAg-positive chronic hepatitis B showing response to 24-week clevudine therapy.
PloS one (2012-12-05)
Hwi Young Kim, Hee-Dae Kwon, Tae Soo Jang, Jisun Lim, Hyo-Suk Lee
ABSTRACT
Modeling of short-term viral dynamics of hepatitis B with traditional biphasic model might be insufficient to explain long-term viral dynamics. The aim was to develop a novel method of mathematical modeling to shed light on the dissociation between early and long-term dynamics in previous studies. We investigated the viral decay pattern in 50 patients from the phase III clinical trial of 24-week clevudine therapy, who showed virological response and HBsAg decline. Immune effectors were added as a new compartment in the model equations. We determined some parameter values in the model using the non-linear least square minimization method. Median baseline viral load was 8.526 Log(10)copies/mL, and on-treatment viral load decline was 5.683 Log(10)copies/mL. The median half-life of free virus was 24.89 hours. The median half-life of infected hepatocytes was 7.39 days. The viral decay patterns were visualized as triphasic curves with decreasing slopes over time: fastest decay in the first phase; slowest in the third phase; the second phase in between. In the present study, mathematical modeling of hepatitis B in patients with virological response and HBsAg decline during 24-week antiviral therapy showed triphasic viral dynamics with direct introduction of immune effectors as a new compartment, which was thought to reflect the reduction of clearance rate of infected cells over time. This modeling method seems more appropriate to describe long-term viral dynamics compared to the biphasic model, and needs further validation.
Sigma-Aldrich
Uracil 1-β-D-arabinofuranoside, lymphoma antiproliferative